Key Documents
Z719641
Sorenson low binding standard tips
maximum volume 300 μL, rack of 96, sterile
Select a Size
960 EA
$226.00
4800 EA
$823.00
$226.00
Estimated to ship onApril 14, 2025
Select a Size
Change View
960 EA
$226.00
4800 EA
$823.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
300 μL
sterility:
sterile
feature:
barrier: no
graduations
packaging:
case of 4800 ea (50 racks of 96 tips)
rack of 96
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Sorenson 31790T
$226.00
Estimated to ship onApril 14, 2025
material
colorless
polypropylene
sterility
sterile
feature
barrier: no
graduations
packaging
case of 4800 ea (50 racks of 96 tips)
rack of 96
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sorenson 31790T
maximum volume
300 μL
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)