CLS4418
Corning® 8-cap strips for 1.2 mL cluster tubes and cluster tube strips
96 well cluster tube 8-cap strips, polyethylene, sterile, 120/cs
Synonym(s):
cap strips, tube caps
About This Item
Recommended Products
material
polyethylene
sterility
sterile
feature
Plate feature: 8-cap strips
Plate format: 96 well cluster tube
packaging
pack of 12
case of 120 (10 × 12 strips, pkg quantity/case)
manufacturer/tradename
Corning 4418
color
clear
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service