Skip to Content
Merck
All Photos(1)

Key Documents

Z709247

ART® self-sealing barrier pipette tips

ART 200U, volume range 1-200 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$303.00
5 PKG
$1,070.00

$303.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 PKG
$303.00
5 PKG
$1,070.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
1—200 μL
sterility:
sterile
feature:
barrier
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2770

$303.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

colorless tip

sterility

sterile

feature

barrier

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2770

technique(s)

PCR: suitable

maximum volume

200 μL

volume range

1-200 μL

volume

1—200 μL

compatibility

for use with

  • Capp Aero 200 μl
  • Eppendorf Titerman 4908 30-300 μl
  • Finnpipette 4027/4500 5-40 μl
  • Finnpipette Multi-Channel (Limited Draw) 50-300 μl
  • Pipetman P-200 20-200 μl
  • Rainin EDP 2 20-250 μl
  • Rainin EDP Plus 20-250 μl
  • Rainin Pipet Plus 20-200 μl
  • Rainin Pipetman 20-200 μl
  • Transferpette Multi-Channel 20-200 μl

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

SoftFit-L tip design and sturdy rack are ideal for multi-channel pipettors

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).[1][2][3]

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service