Skip to Content
Merck
All Photos(1)

Key Documents

Z569763

ART® self-sealing barrier pipette tips

ART 20P, volume range 0.5-20 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

960 EA
$323.00
4800 EA
$1,200.00

$323.00


Estimated to ship onMay 30, 2025



Select a Size

Change View
960 EA
$323.00
4800 EA
$1,200.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
0.5—20 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 960 ea
manufacturer/tradename:
Molecular BioProducts 2149P

$323.00


Estimated to ship onMay 30, 2025


material

colorless
colorless tip

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 960 ea

manufacturer/tradename

Molecular BioProducts 2149P

technique(s)

PCR: suitable

maximum volume

20 μL

volume range

0.5-20 μL

volume

0.5—20 μL

Looking for similar products? Visit Product Comparison Guide

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service