Key Documents
Z659339
Bandelin SONOPULS ultrasonic homogenizer accessories
Flow-through vessel DG 5 for HD 2200 homogenizer
Synonym(s):
sonification vessel
Select a Size
1 EA
$513.00
$513.00
Estimated to ship onApril 09, 2025
Select a Size
Change View
1 EA
$513.00
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
$513.00
Estimated to ship onApril 09, 2025
material
glass
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 482
availability
available only in EU
volume × I.D. × H
80 mL × 35 mm × 95 mm
General description
Flow vessel DG with cooling jacket, for continuous sonication of larger quantities in the flow. The cooling jacket allows temperature control using liquid coolants during sonication via existing hose connections.
Features and Benefits
  • Simultaneous sound and cooling
  • Optimal process observation
Choose from one of the most recent versions:
Certificates of Analysis (COA)