Key Documents
Z655422
Hanna rugged platinium & gold tips ORP electrodes for general use
Gold pin tip - reduction reactions
About This Item
UNSPSC Code:
41115603
manufacturer/tradename
Hanna HI 4430B
parameter
0-80 °C temp. range
2 bar max. pressure
General description
  • Single, Ag/AgCI
  • Ceramic, single junction
  • Electrolyte - gel
  • 1 m coaxial cable
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Choose from one of the most recent versions:
Certificates of Analysis (COA)