Skip to Content
Merck
All Photos(1)

Key Documents

1.02703

Supelco

Copper

fine powder particle size < 63 µm (> 230 mesh ASTM) EMSURE®

Synonym(s):

Copper

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
12141711
EC Index Number:
231-159-6
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥99% (complexometric)

form

solid

impurities

≤0.02% Substances insoluble in nitric acid

mp

1083 °C

density

8.96 g/cm3 at 20 °C

bulk density

1290 kg/m3

cation traces

Ag: ≤0.002%
As: ≤0.0005%
Fe: ≤0.005%
Mn: ≤0.001%
P: ≤0.001%
Pb: ≤0.01%
Sb: ≤0.001%
Sn: ≤0.01%

storage temp.

2-30°C

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

Related Categories

Linkage

Replaces: CX1925-4; CX1925-2; CX1925

Analysis Note

Assay (complexometric): ≥ 99 %
Substances insoluble in nitric acid: ≤ 0.02 %
P (phosphorus): ≤ 0.001 %
Ag (Silver): ≤ 0.002 %
As (Arsenic): ≤ 0.0005 %
Fe (Iron): ≤ 0.005 %
Mn (Manganese): ≤ 0.001 %
Pb (Lead): ≤ 0.01 %
Sb (Antimony): ≤ 0.001 %
Sn (Tin): ≤ 0.01 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service