Skip to Content
Merck
All Photos(1)

Key Documents

Z637394

Mucocit-P disinfecting powder cleaner

weight 2.5 kg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800

manufacturer/tradename

BRAND 44920

weight

2.5 kg

General description

Powerful and gentle to labware. Free from phosphates, aldehydes and phenols. The solution has a pleasant fresh fragrance. For disinfection of instruments.

Microbiological efficiency / concentration for use:
  • Bacteria (inc TbB) and fungi to DGHM: 3% (30 g/L)/1 hour
  • Limited virucidal efficacy (incl. HIV, HBV, HCV: 1% (10 g/L)/5 min.

Other Notes

BR44920 (India)

Legal Information

Mucocit is a trademark of Merz GmbH & Co. KG

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service