Key Documents
Z637149
BRAND® UV cuvette lids, PE
round orange lid
Select a Size
100 EA
$44.70
$44.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
100 EA
$44.70
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
$44.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
round orange lid
manufacturer/tradename
BRAND
General description
Round covers allow a tight closure and make it possible to store samples down to -20°C.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)