Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
UV cuvette lids, PE
Products
Product Category
lids (4)
Manufacturer
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z637117
BRAND
®
UV cuvette lids, PE,
round blue lid
Expand
View Pricing
Z637122
BRAND
®
UV cuvette lids, PE,
round yellow lid
Expand
View Pricing
Z637149
BRAND
®
UV cuvette lids, PE,
round orange lid
Expand
View Pricing
Z637130
BRAND
®
UV cuvette lids, PE,
round green lid
Expand
View Pricing