Skip to Content
Merck
All Photos(1)

Key Documents

Z614246

Vivaspin 2 centrifugal concentrators

MWCO 30 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$309.00

$309.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
25 EA
$309.00

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

$309.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polycarbonate housing
polyethersulfone membrane

sterility

non-sterile

feature

holdup volume 10 μL

packaging

package of 25 ea

manufacturer/tradename

Sartorius VS0221

parameter

8000 × g max. centrifugal force

dead stop volume

8 μL

membrane area

1.2 cm2

tube size

~15 mL

working volume

2 mL

pore size

30 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

The Vivaspin 2 combines the speed of the classic Vivaspin products with low internal surface and membrane area for superior recoveries from very dilute solutions. Unique to the Vivaspin 2 is the choice of directly pipetting the concentrate from the dead stop pocket built into the bottom of the concentrator, or alternatively reverse spinning into the concentrate recovery cap which can then be sealed for storage. Both methods result in near total concentrate recoveries.

  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service