Merck
  • LAMTOR/Ragulator is a negative regulator of Arl8b- and BORC-dependent late endosomal positioning.
LAMTOR/Ragulator is a negative regulator of Arl8b- and BORC-dependent late endosomal positioning.
The Journal of cell biology (2017-10-11)
Przemyslaw A Filipek, Mariana E G de Araujo, Georg F Vogel, Cedric H De Smet, Daniela Eberharter, Manuele Rebsamen, Elena L Rudashevskaya, Leopold Kremser, Teodor Yordanov, Philipp Tschaikner, Barbara G Fürnrohr, Stefan Lechner, Theresia Dunzendorfer-Matt, Klaus Scheffzek, Keiryn L Bennett, Giulio Superti-Furga, Herbert H Lindner, Taras Stasyk, Lukas A Huber
ABSTRACT
Signaling from lysosomes controls cellular clearance and energy metabolism. Lysosomal malfunction has been implicated in several pathologies, including neurodegeneration, cancer, infection, immunodeficiency, and obesity. Interestingly, many functions are dependent on the organelle position. Lysosomal motility requires the integration of extracellular and intracellular signals that converge on a competition between motor proteins that ultimately control lysosomal movement on microtubules. Here, we identify a novel upstream control mechanism of Arl8b-dependent lysosomal movement toward the periphery of the cell. We show that the C-terminal domain of lyspersin, a subunit of BLOC-1-related complex (BORC), is essential and sufficient for BORC-dependent recruitment of Arl8b to lysosomes. In addition, we establish lyspersin as the linker between BORC and late endosomal/lysosomal adaptor and mitogen activated protein kinase and mechanistic target of rapamycin activator (LAMTOR) complexes and show that epidermal growth factor stimulation decreases LAMTOR/BORC association, thereby promoting BORC- and Arl8b-dependent lysosomal centrifugal transport.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Indole-3-butyric acid potassium salt, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vivaspin 500 centrifugal concentrators, MWCO 5 kDa
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Leupeptin, synthetic, ≥85% (HPLC)
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture