Z375357
Bright-Line™ Hemacytometer replacement cover slip
replacement cover slip for the Bright-Line Hemacytometer: Z359629
Synonym(s):
Microscope Cover Glass
All Photos(1)
About This Item
Recommended Products
material
glass
packaging
pkg of 1 ea
binding type
non-treated surface
General description
This is the replacement cover slip for the Bright-Line Hemacytometer, Prod. No. Z359629
This product consists of a single cover slip. The microscope slide cover slips are polished soda lime glass, they are rectangular in shape and measure: 20 mm x 26 mm x 0.5 mm thick.
This product consists of a single cover slip. The microscope slide cover slips are polished soda lime glass, they are rectangular in shape and measure: 20 mm x 26 mm x 0.5 mm thick.
Legal Information
Bright-Line is a trademark of Cambridge Instruments, Inc.
Articles
Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.