Skip to Content
Merck

Skip To

C8181

Cover glasses

size 24 mm × 50 mm

Synonym(s):

Glass cover slips, Microscope cover slips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 pkg
Check Cart for Availability
$27.90
1 case
Check Cart for Availability
$403.00

About This Item

NACRES:
MA.01
UNSPSC Code:
41122600
Packaging:
case of 10 1-oz pkgs, pkg of ~65 per ounce

$27.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

No. 1 thickness (0.13-0.16 mm)

packaging

case of 10 1-oz pkgs, pkg of ~65 per ounce

size

24 mm × 50 mm


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C9056C7931BR470819
description

size 24 mm × 50 mm

description

size 24 mm × 60 mm

description

size 24 mm × 40 mm

description

L × W 24 mm × 50 mm, rectangular

packaging

case of 10 1-oz pkgs, pkg of ~65 per ounce

packaging

pkg of ~54 per ounce, case of 10 1-oz pkgs

packaging

case of 10 1-oz pkgs, pkg of ~83 per ounce

packaging

-

size

24 mm × 50 mm

size

24 mm × 60 mm

size

24 mm × 40 mm

size

-

description

No. 1 thickness (0.13-0.16 mm)

description

No. 1 thickness (0.13-0.16 mm)

description

No. 1 thickness (0.13-0.16 mm)

description

-


Still not finding the right product?

Explore all of our products under Cover glasses




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library