Skip to Content
Merck
All Photos(1)

Key Documents

Z370665

Sigma-Aldrich

Biohazard waste container

capacity 19 L (5 gal)

Synonym(s):

autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$409.00

$409.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
1 EA
$409.00

About This Item

UNSPSC Code:
41123300
NACRES:
NB.35

$409.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


packaging

pack of 1 ea

manufacturer/tradename

Nalgene 6370-0005

O.D. × H

28 cm (11 in.) × 43 cm (17 in.)

capacity

19 L (5 gal)

Looking for similar products? Visit Product Comparison Guide

General description

Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials.

Convenient one-step sterilization; no need to remove bag. Port opening in cover allows one-handed disposal of materials; limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust, dent or puncture.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this autoclavable????

    1 answer
    1. Yes, this product is specifically cited by the manufacturer Nalgene as being suitable for autoclaving bio-hazardous waste.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service