Key Documents
Z281344
Laboratory brushes
Laboratory brushes
Flask, L 12 in., brush diam. × L 1 3/8 in. × 3 1/2 in.
Select a Size
4 EA
$65.00
$65.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
4 EA
$65.00
About This Item
UNSPSC Code:
47131605
NACRES:
NB.35
$65.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 4 ea
L
12 in.
brush diam. × L
1 3/8 in. × 3 1/2 in.
General description
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Buonasera vorrei sapere se laboratory brushes Cod. Z281344 il filo che regge le setole in nylon sono in acciaio inox. Grazie
    1 answer
    1. Yes, it is 304 stainless steel.
      Helpful?
Reviews
Active Filters
  1. BFN
    • Review 1
    • Votes 0
    4 out of 5 stars.
    Different
    What I got is very different to what I sought as shown in the picture above.
    Helpful?
    1. Response from MilliporeSigma:
      Thank you for your review. We have updated the primary image on this page to reflect the type of brush you should be receiving. If you have any further issues using this product please call or email us for additional support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.