Skip to Content
Merck
All Photos(1)

Key Documents

Z367982

Sigma-Aldrich

Cellulose acetate membrane

dialyser capacity 10-200 μL, MWCO 500 Da, diam. 11 mm

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$278.00

$278.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
25 EA
$278.00

About This Item

UNSPSC Code:
41123100
NACRES:
NB.22

$278.00


Estimated to ship onMay 28, 2025


packaging

pkg of 25 membranes

dialyser capacity

10-200 μL

diam.

11 mm

pore size

500 Da MWCO

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Made of cellulose acetate for low protein binding. Nominal molecular weight limit (MWCO) is approximate upper limit of molecular weight to which membrane is permeable.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service