Key Documents
Z362891
Sigma-Aldrich
Nalgene® benchtop β waste container
capacity 2,000 mL
Synonym(s):
lab waste receptical, liquid waste holder
Select a Size
1 EA
$569.00
$569.00
Estimated to ship onApril 08, 2025
Select a Size
Change View
1 EA
$569.00
About This Item
UNSPSC Code:
26142300
NACRES:
NB.35
$569.00
Estimated to ship onApril 08, 2025
material
acrylic
manufacturer/tradename
Nalgene®
capacity
2,000 mL
Related Categories
General description
3/8 in. thick, encases waste bottle without obstructing access. Shield cap fits securely with bottle closure in place. Provided with wide-mouth polyethylene bottle suitable for aqueous solution. Disposal of organic solvents may require a different inner bottle.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)