Key Documents
Z360422
Desiccator plate
diam. 230 mm, 24 holes
Synonym(s):
perforated plate, sample support plate
Select a Size
1 EA
$251.00
6 EA
$1,120.00
$251.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$251.00
6 EA
$1,120.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$251.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
Nalgene® 5312-0230
O.D.
230 mm
diam.
230 mm
General description
This desiccator plate has a greater resistance to thermal shock than porcelain plates. It has a fire-polished surface, and is a light green ceramic bonded to a metal core for inertness and corrosion-resistance. Numbered quadrants are marked for easy location of crucibles, weighing bottles, and other containers. Recommended use with Nalgene desiccator such as D2547, Z354074 and D2797.
The center hole is 7/8 in. diam. The plates have 24 holes.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)