Key Documents
Z360287
Sigma-Aldrich
Carboy spigots
polypropylene
Synonym(s):
plastic spigot, plastic spigots
Select a Size
1 EA
$96.90
$96.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$96.90
About This Item
UNSPSC Code:
41121820
NACRES:
NB.35
$96.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 6422-0010
Related Categories
General description
Adapter accepts 1/4 to 5/16 in. tubing. This spigot is the standard fitting for all Nalgene carboys. Do not autoclave adapter.
Spigots fit all Nalgene carboys. Both are autoclavable when installed on autoclavable carboys. Thread is 11/8 in. - 12 straight female threads. Spout accepts 5/8 in. I.D. tubing.
Choose from one of the most recent versions:
Certificates of Analysis (COA)