Skip to Content
Merck
All Photos(1)

Documents

Y3127

Millipore

Yeast malt agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

composition

Agar, 20.00 g/L
Dextrose, 10.00 g/L
Malt extract, 3.00 g/L
Peptic digest of animal tissue, 5.00 g/L
Yeast extract, 3.00 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

6.2±0.2 (25 °C)

application(s)

agriculture
bioburden testing
clinical testing
environmental
food and beverages

microbiology

suitability

nonselective for Aspergillus spp.
nonselective for Candida spp.
nonselective for Lactobacillus spp.
nonselective for Penicillium spp.
nonselective for Pichia spp.
nonselective for Saccharomyces spp.
nonselective for Zygosaccharomyces spp.
nonselective for aciduric bacteria
nonselective for molds (General Media)
nonselective for yeasts (General Media)

Related Categories

Application

Used for the isolation and cultivation of yeasts, molds and other aciduric microorganisms.

Preparation Note

Suspend 41 grams of Yeast Malt Agar in 1000 ml of distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 121 °C for 15 minutes. For preparing a selective media acidify the media to pH 3.0-4.0 or add antibiotics. Do not heat the media after the addition of acid.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service