Skip to Content
Merck
All Photos(1)

Key Documents

Y1396

Sigma-Aldrich

YFLLRNP trifluoroacetate

≥98%

Synonym(s):

L-Tyrosyl-L-phenylalanyl-L-leucyl-L-leucyl-L-arginyl-L-asparaginyl-L-Proline, Tyr-Phe-Leu-Leu-Arg-Asn-Pro, YFLLRNP-OH

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$235.00

$235.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$235.00

About This Item

Empirical Formula (Hill Notation):
C45H67N11O10 · xC2HF3O2
Molecular Weight:
922.08 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

$235.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Quality Level

Assay

≥98%

form

solid

solubility

deionized water: 1 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(N)=O)C(=O)N3CCC[C@H]3C(O)=O

InChI

1S/C45H67N11O10.C2HF3O2/c1-25(2)20-32(40(61)51-31(12-8-18-50-45(48)49)39(60)55-35(24-37(47)58)43(64)56-19-9-13-36(56)44(65)66)53-41(62)33(21-26(3)4)54-42(63)34(23-27-10-6-5-7-11-27)52-38(59)30(46)22-28-14-16-29(57)17-15-28;3-2(4,5)1(6)7/h5-7,10-11,14-17,25-26,30-36,57H,8-9,12-13,18-24,46H2,1-4H3,(H2,47,58)(H,51,61)(H,52,59)(H,53,62)(H,54,63)(H,55,60)(H,65,66)(H4,48,49,50);(H,6,7)/t30-,31-,32-,33-,34-,35-,36-;/m0./s1

InChI key

HMDRRCMAXMJXKF-WDYYMHPRSA-N

Application

YFLLRNP trifluoroacetate may be used as a powerful tool for differentiating the numerous possible activation states of the platelet thrombin receptor.

Biochem/physiol Actions

YFLLRNP is a Galpha12/13-stimulating peptide that activates the alpha granule secretion due to costimulation with either ADP or serotonin/epinephrine.[1]
YFLLRNP is a PAR-1-specific partial agonist.
YFLLRNP is a PAR-1-specific partial agonist. The compound selectively activates the G(12/13) signaling cascade resulting in platelet shape.

Preparation Note

YFLLRNP trifluoroacetate dissolves in deionized water at 1 mg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M Quinton et al.
Journal of thrombosis and haemostasis : JTH, 2(6), 978-984 (2004-05-14)
Alpha granule release plays an important role in propagating a hemostatic response upon platelet activation. We evaluated the ability of various agonists to cause alpha granule release in platelets. Alpha granule release was measured by determining P-selectin surface expression in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service