Skip to Content
Merck
All Photos(4)

Key Documents

N4505

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide, reduced dipotassium salt

Synonym(s):

β-DPNH, β-NADH, DPNH, Diphosphopyridine nucleotide, reduced form, NADH

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$272.00
500 MG
$458.00
1 G
$848.00

$272.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$272.00
500 MG
$458.00
1 G
$848.00

About This Item

Linear Formula:
C21H27N7O14P2K2
CAS Number:
Molecular Weight:
741.62
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

$272.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

biological source

synthetic

Assay

≥95%

form

powder

solubility

10 mM NaOH: soluble-100 mg/mL, clear to hazy, faintly yellow to yellow

storage temp.

−20°C

SMILES string

[K].NC(=O)C1=CN(C=CC1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)n4cnc5c(N)ncnc45)C(O)C2O

InChI

1S/C21H29N7O14P2.K.H/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;

InChI key

KMRPNNQOKAHXEZ-UHFFFAOYSA-N

Gene Information

human ... IMPDH2(3615)

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).

Biochem/physiol Actions

Electron donor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luisa Schwarzmann et al.
Bioscience reports, 41(1) (2021-01-05)
Nicotinamide adenine dinucleotide (NAD) is a coenzyme in metabolic reactions and cosubstrate in signaling pathways of cells. While the intracellular function of NAD is well described, much less is known about its importance as an extracellular molecule. Moreover, there is
Cornelia Wetzker et al.
Scientific reports, 9(1), 19534-19534 (2019-12-22)
Metabolic profiles vary across all levels of biological diversity, from cells to taxa. Two-photon fluorescence lifetime imaging microscopy (FLIM) facilitates metabolic characterisation of biological specimens by assaying the intrinsic autofluorescence of the ubiquitous coenzymes NAD(P)H and FAD. The potential of
Sritama De Sarkar et al.
Parasitology research, 118(1), 335-345 (2018-11-25)
Berberine chloride, a plant-derived isoquinoline alkaloid, has been demonstrated to have leishmanicidal activity, which is mediated by generation of a redox imbalance and depolarization of the mitochondrial membrane, resulting in a caspase-independent apoptotic-like cell death. However, its impact on mitochondrial
Erika Chacin et al.
Nature communications, 12(1), 5224-5224 (2021-09-03)
The replication of chromosomes during S phase is critical for cellular and organismal function. Replicative stress can result in genome instability, which is a major driver of cancer. Yet how chromatin is made accessible during eukaryotic DNA synthesis is poorly
Peter M Fernandes et al.
The FEBS journal, 287(13), 2847-2861 (2019-12-16)
Trypanosomatids possess glycosome organelles that contain much of the glycolytic machinery, including phosphofructokinase (PFK). We present kinetic and structural data for PFK from three human pathogenic trypanosomatids, illustrating intriguing differences that may reflect evolutionary adaptations to differing ecological niches. The

Questions

1–4 of 4 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the extinction coefficient for Product N4505, β-Nicotinamide adenine dinucleotide, reduced dipotassium salt?

    1 answer
    1. EmM = 6.22 (340 nm) and 14.4 (259 nm, pH 9.5)

      Helpful?

  3. What information do you have on the solubility and solution stability of Product N4505, β-Nicotinamide adenine dinucleotide, reduced dipotassium salt?

    1 answer
    1. Solubility and solution stability information can be found on the product information sheet (under Documents, above).

      Helpful?

  4. What is the absorption maximum and the fluorescence emission maximum wavelengths for Product N4505, β-Nicotinamide adenine dinucleotide, reduced dipotassium salt?

    1 answer
    1. The absorption maximum for NADH is at 340 nm and the fluorescence emission maximum is at 460 nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service