Skip to Content
Merck
All Photos(1)

Key Documents

WPB01423WA

Whirl-Pak® Pre-moistened Speci-Sponge® Bag

with Sterile Glove

Synonym(s):

sampling bag, surface sampling bag

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$458.00

$458.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 EA
$458.00

About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

$458.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

sterility

sterile; γ-irradiated

packaging

pkg of 100 ea

manufacturer/tradename

Nasco B01423WA

W × L × thickness

11.5 cm × 23 cm × 0.064 mm

capacity

532 mL × 18 oz
532 mL capacity(18 oz)

Looking for similar products? Visit Product Comparison Guide

General description

Environmental surface sampling can be faster and easier by using the Whirl-Pak pre-moistened Speci-Sponge bag. The sponge is ready to use and is hydrated with 10 ml of neutralizing buffer that will neutralize several surface sanitizers including quaternary ammonium compounds, phenolics, iodine preps, chlorine preps, mercurials, formaldehyde, and glutaraldehyde. To use, squeeze out excess buffer from the sponge - this can be done by squeezing the sponge from the outside with two hands or squeezing the sponge with a sterile gloved hand after opening the bag. Wipe the surface to be tested, return the sponge to the bag, close, and send to the lab for analysis.

The buffer has a 24 month shelf life from date of manufacture and each carton is marked with an expiration date; no refrigeration is necessary. The bags are packaged in a foil liner bag to prevent evaporation of the buffer. Buffer contains monopotassium phosphate, sodium thiosulfate, and aryl sulfonate complex.

Legal Information

Speci-Sponge is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service