Skip to Content
Merck
All Photos(1)

Key Documents

Z699462

Dilution bottle

Butterfield′s Buffer, pack of 72 × 99 mL

Synonym(s):

dilution bottle, dilution bottles, phosphate buffer

Sign Into View Organizational & Contract Pricing

Select a Size

72 EA
$367.00

$367.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
72 EA
$367.00

About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

$367.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pack of 72 × 99 mL

manufacturer/tradename

Maine Mfg 10498503

Looking for similar products? Visit Product Comparison Guide

General description

Butterfield′s Phosphate Buffer contains monobasic potassium phosphate and is used extensively in the food, dairy and pharmaceutical industries. Offered in 90 mL and 99 mL volumes for easy 1:10 and 1:100 dilutions. It is recommended as a general diluent in laboratory procedures by the Federal Drug Administration and in the Bacteriological Analytical Manual. This product is prepared according to Standard Methods for the Examination of Water and Wastewater for use in water testing.
Whatman sterile dilution bottles are designed for sample dilution of water, dairy products, foods, and pharmaceuticals prior to microbiological testing. Final pH for all solutions is 7.2 pH ± 0.2 pH at 25°C. The buffers are supplied in an easy open, flip-top, plastic container with a tamper-evident seal.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service