Skip to Content
Merck
All Photos(6)

Documents

WH0008192M1

Sigma-Aldrich

Monoclonal Anti-CLPP antibody produced in mouse

clone 300, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog (E. coli)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

300, monoclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLPP(8192)

General description

CLPP (caseinolytic mitochondrial matrix peptidase proteolytic subunit) protein contains an aqueous chamber formed by face-to-face assembly of the two heptameric rings that have proteolytic active sites within. It is a member of the peptidase family S14. The CLPP gene is localized to human chromosome 19p13.3.

Immunogen

CLPP (NP_006003, 178 a.a. ~ 277 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
HQPSGGARGQATDIAIQAEEIMKLKKQLYNIYAKHTKQSLQVIESAMERDRYMSPMEAQEFGILDKVLVHPPQDGEDEPTLVQKEPVEAAPAAEPVPAST

Application

Monoclonal Anti-CLPP antibody produced in mouse has been used in Western Blotting.

Biochem/physiol Actions

CLPP (caseinolytic mitochondrial matrix peptidase proteolytic subunit) gene encodes a protease component of the Clp complex that hydrolyzes peptides and various proteins in the presence of ATP. The protein requires magnesium for its activity. CLPP is found to be associated with the inner mitochondrial membrane.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallography and mutagenesis point to an essential role for the N-terminus of human mitochondrial ClpP
Sung Gyun Kang
Journal of Structural Biology, 148 (2004)
Gavin Pharaoh et al.
Redox biology, 8, 430-438 (2016-05-22)
Mice deficient in the electron transport chain (ETC) complex IV assembly protein SURF1 have reduced assembly and activity of cytochrome c oxidase that is associated with an upregulation of components of the mitochondrial unfolded protein response (UPR(MT)) and increased mitochondrial
Perrault syndrome is caused by recessive mutations in CLPP, encoding a mitochondrial ATP-dependent chambered protease
Emma M Jenkinson
American Journal of Human Genetics, 92 (2013)
Ablation of the mitochondrial complex IV assembly protein Surf1 leads to increased expression of the UPRMT and increased resistance to oxidative stress in primary cultures of fibroblasts
Gavin Pharaoh
Redox Biology (2016)
Shylesh Bhaskaran et al.
EMBO reports, 19(3) (2018-02-09)
Caseinolytic peptidase P (ClpP) is a mammalian quality control protease that is proposed to play an important role in the initiation of the mitochondrial unfolded protein response (UPRmt), a retrograde signaling response that helps to maintain mitochondrial protein homeostasis. Mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service