Skip to Content
Merck
All Photos(1)

Documents

AB16530

Sigma-Aldrich

Anti-BAFF Antibody, CT

Chemicon®, from rabbit

Synonym(s):

TALL-1, THANK, BLyS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated BAFF (for B cell Activating Factor belonging to the TNF Family), BlyS (for B Lymphocyte Stimulator), TALL-1 (for TNF- and ApoL-related Leukocyte-expressed Ligand), and THANK (for TNF Homologue that Activate Apoptosis, NF-kB and c-jun N-terminal kinase) (Moore et al. 1999; Schneider et al. 1999; Shu et al. 1999; Mukhopadhyay et al. 1999). BAFF/BlyS was characterized as a B cell stimulator since it induced B cell proliferation and immunoglobulin secretion (Moore et al. 1999; Schneider et al. 1999). Two receptors for BAFF were recently identified and designated TACI and BCMA (Gross et al. 2000). BAFF and its receptors are involved in the development of systemic lupus erythaematosus and other B cell associated autoimmune diseases (Gross et al. 2000; Khare et al. 2000). Like TNFalpha and TRAIL, THANK was shown to activate NF-kB and c-jun N-terminal kinase (JNK) and to induce apoptosis (Mukhopadhyay et al. 1999).

Immunogen

Epitope: C-terminus
Rabbit anti-BAFF (C-term) polyclonal antibody was raised against a peptide corresponding to amino acids 254 to 269 of human BAFF (Moore et al. 1999; Schneider et al. 1999; Shu et al. 1999; Mukhopadhyay et al. 1999).

Application

Anti-BAFF Antibody, C-terminus detects level of BAFF & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
Western blot: 0.25 to 1 μg/mL.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Madayag et al.
Immunity, inflammation and disease, 10(11), e729-e729 (2022-10-28)
Epstein-Barr virus (EBV) establishes a lifelong infection in human B cells where the virus consistently expresses Latent Membrane Protein 2A (LMP2A) to promote B cell survival. A prior study indicates that LMP2A may increase the production of the pro-survival factor
Pleiotropic effects of BAFF on the senescence-associated secretome and growth arrest.
Rossi, et al.
eLife, 12 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service