Skip to Content
Merck
All Photos(1)

Key Documents

V0750

Sigma-Aldrich

L-Valine benzyl ester hydrochloride

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2 · HCl
CAS Number:
Molecular Weight:
243.73
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Valine benzyl ester hydrochloride,

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

Cl.CC(C)[C@H](N)C(=O)OCc1ccccc1

InChI

1S/C12H17NO2.ClH/c1-9(2)11(13)12(14)15-8-10-6-4-3-5-7-10;/h3-7,9,11H,8,13H2,1-2H3;1H/t11-;/m0./s1

InChI key

ZIUNABFUHGBCMF-MERQFXBCSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dawei Ma et al.
Organic letters, 4(14), 2377-2380 (2002-07-06)
[reaction: see text] Condensation of L-valine benzyl ester toluenesulfonic acid salt with a substituted cyclohexadione followed by aromatization with the assistance of NBS provides an N-aryl L-valine benzyl ester. This intermediate is converted into 7-substituted benzolactam-V8s using an asymmetric Strecker

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service