Skip to Content
Merck
All Photos(1)

Key Documents

364517

Sigma-Aldrich

L-Tryptophan methyl ester hydrochloride

98%, for peptide synthesis

Synonym(s):

(S)-Tryptophan methyl ester hydrochloride, H-(L)-Trp-OMe hydrochloride, L-(-)-Tryptophan methyl ester hydrochloride, Tryptophan methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2 · HCl
CAS Number:
Molecular Weight:
254.71
Beilstein:
4240280
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

L-Tryptophan methyl ester hydrochloride, 98%

Assay

98%

optical activity

[α]20/D +18°, c = 5 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

218-220 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)[C@@H](N)Cc1c[nH]c2ccccc12

InChI

1S/C12H14N2O2.ClH/c1-16-12(15)10(13)6-8-7-14-11-5-3-2-4-9(8)11;/h2-5,7,10,14H,6,13H2,1H3;1H/t10-;/m0./s1

InChI key

XNFNGGQRDXFYMM-PPHPATTJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Tryptophan methyl ester hydrochloride can be used as a reactant to prepare:
  • Oxamoyl derivatives of N-(2-aminobenzoyl)-L-tryptophan (dipeptides) by acylation reaction with 3,1-benzoxazinones.[1]
  • Tadalafil (Cialis), a type-V phosphodiesterase (PDE5) inhibitor.[2]
  • Isoroquefortine C.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno M Schiavi et al.
The Journal of organic chemistry, 67(3), 620-624 (2002-02-22)
A short and efficient total synthesis of isoroquefortine C, the 3,12-(Z)-isomer of roquefortine C, from L-tryptophan methyl ester hydrochloride and 4(5)-(hydroxy)methylimidazole hydrochloride is described.
Christine Böttcher et al.
Journal of experimental botany, 62(12), 4267-4280 (2011-05-06)
Nine Gretchen Hagen (GH3) genes were identified in grapevine (Vitis vinifera L.) and six of these were predicted on the basis of protein sequence similarity to act as indole-3-acetic acid (IAA)-amido synthetases. The activity of these enzymes is thought to
Synthesis of acyl derivatives of L-tryptophan containing residues of anthranilic and oxalic acids
Shemchuk LA, et al.
Russ. J. Org. Chem., 44(5), 693-696 (2008)
Synthesis of tadalafil (Cialis) from L-tryptophan
Xiao Sen, et al.
Tetrahedron Asymmetry, 20(18), 2090-2096 (2009)
Christopher Jonathan Daraius Austin et al.
Amino acids, 39(2), 565-578 (2010-02-09)
The first step in the kynurenine pathway of tryptophan catabolism is the cleavage of the 2,3-double bond of the indole ring of tryptophan. In mammals, this reaction is performed independently by indoleamine 2,3-dioxygenase-1 (IDO1), tryptophan 2,3-dioxygenase (TDO) and the recently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service