Skip to Content
Merck
All Photos(1)

Key Documents

V0131

Sigma-Aldrich

Vasoactive Intestinal Peptide Fragment 1-12 human, porcine, rat

≥97% (HPLC)

Synonym(s):

VIP 1-12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H88N18O22
CAS Number:
Molecular Weight:
1425.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Product Name

Vasoactive Intestinal Peptide Fragment 1-12 human, porcine, rat, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

solid

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(C)NC(=O)C(CC(O)=O)NC(=O)C(CO)NC(=O)C(N)Cc1cnc[nH]1)C(=O)NC(Cc2ccccc2)C(=O)NC(C(C)O)C(=O)NC(CC(O)=O)C(=O)NC(CC(N)=O)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(C(C)O)C(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C61H88N18O22/c1-27(2)46(77-49(89)28(3)69-51(91)40(22-44(85)86)73-56(96)42(25-80)76-50(90)35(62)20-33-24-66-26-68-33)57(97)74-38(18-31-10-7-6-8-11-31)55(95)79-48(30(5)82)59(99)75-41(23-45(87)88)53(93)72-39(21-43(63)84)52(92)71-37(19-32-13-15-34(83)16-14-32)54(94)78-47(29(4)81)58(98)70-36(60(100)101)12-9-17-67-61(64)65/h6-8,10-11,13-16,24,26-30,35-42,46-48,80-83H,9,12,17-23,25,62H2,1-5H3,(H2,63,84)(H,66,68)(H,69,91)(H,70,98)(H,71,92)(H,72,93)(H,73,96)(H,74,97)(H,75,99)(H,76,90)(H,77,89)(H,78,94)(H,79,95)(H,85,86)(H,87,88)(H,100,101)(H4,64,65,67)

InChI key

OZQVVUDUPMJWPH-UHFFFAOYSA-N

Gene Information

human ... VIP(7432)

Amino Acid Sequence

His-Ser-Asp-Ala-Val-Phe-Thr-Asp-Asn-Tyr-Thr-Arg

Biochem/physiol Actions

Ligand for CD4 receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Sung et al.
Neuropeptides, 33(6), 435-446 (2000-02-05)
It is well known that psoriasis, an immunogenetic cutaneous disorder whose major pathogenic findings are epidermal hyperplasia and T-cell infiltration, is aggravated by psychological stresses. Although the exact mechanism is not yet clarified, antidromic secretion of neuropeptides by cutaneous nerve
M Ichinose et al.
Regulatory peptides, 54(2-3), 457-466 (1994-12-15)
The effect of VIP on phagocytosis in peritoneal macrophages was examined by means of flow cytometry (FCM). This assay revealed that VIP suppressed phagocytosis in a dose-dependent manner. VIP(1-12) did not suppress phagocytosis. VIP(10-28) was more suppressive than VIP(1-28). A
F Séjourné et al.
The American journal of physiology, 273(1 Pt 2), R287-R292 (1997-07-01)
The purpose of this study was to begin to determine the mechanisms underlying vasodilation elicited by vasoactive intestinal peptide (VIP) in sterically stabilized liposomes (SSL) in the in situ peripheral microcirculation. Using intravital microscopy, we found that suffusion of VIP
S Chakder et al.
The Journal of pharmacology and experimental therapeutics, 266(1), 392-399 (1993-07-01)
Because no significant information exists regarding the structure-activity of vasoactive intestinal polypeptide (VIP) to gut smooth muscle, we performed functional studies in vitro on opossum internal anal sphincter (IAS) smooth muscle strips and supplemented them with binding studies to assess
P Sacerdote et al.
Journal of neuroscience research, 18(1), 102-107 (1987-01-01)
A five-amino-acid (TDNYT) sequence of vasoactive intestinal polypeptide (VIP) shares homology with the proposed attachment sequences of the human immunodeficiency virus (HIV). Synthetic peptides with these sequences have previously been shown to block viral envelope (gp120) binding and HIV infectivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service