Skip to Content
Merck
All Photos(1)

Key Documents

A0911

Sigma-Aldrich

[Ile7]-Angiotensin III acetate salt hydrate

≥95% (HPLC)

Synonym(s):

Arg-Val-Tyr-Ile-His-Pro-Ile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$175.00

$175.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$175.00

About This Item

Empirical Formula (Hill Notation):
C43H68N12O9 · xC2H4O2 · yH2O
Molecular Weight:
897.08 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$175.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Product Name

[Ile7]-Angiotensin III acetate salt hydrate, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

O.CC(O)=O.CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@@H](NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](NC(=O)[C@@H](N)CCCNC(N)=N)C(C)C)[C@@H](C)CC)C(O)=O

InChI

1S/C43H68N12O9.C2H4O2.H2O/c1-7-24(5)34(40(61)51-31(20-27-21-47-22-49-27)41(62)55-18-10-12-32(55)38(59)54-35(42(63)64)25(6)8-2)53-37(58)30(19-26-13-15-28(56)16-14-26)50-39(60)33(23(3)4)52-36(57)29(44)11-9-17-48-43(45)46;1-2(3)4;/h13-16,21-25,29-35,56H,7-12,17-20,44H2,1-6H3,(H,47,49)(H,50,60)(H,51,61)(H,52,57)(H,53,58)(H,54,59)(H,63,64)(H4,45,46,48);1H3,(H,3,4);1H2/t24-,25-,29-,30-,31-,32-,33-,34-,35-;;/m0../s1

InChI key

YXASJLHKVNKIRW-ZPJCNHGSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

In [Ile7]-Angiotensin III (Ile7-AIII) the seventh amino acid of angiotensin III (phe) is replaced with isoleucine. [Ile7]-Angiotensin III (Ile7 AIII) is an angiotensin III (A III) receptor antagonist used to selectively block the action of Angiotensin III.
[Ile7]-Angiotensin III is (Ile7-Ang III) used as a peptide angiotensin receptor antagonist often in comparison with other Ang antagonists such as saralasin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Papouchado et al.
Journal of autonomic pharmacology, 15(1), 1-8 (1995-02-01)
1. Effects of angiotensin III (A III) on 3H-noradrenaline (3H-NA) total, neuronal and non-neuronal uptake, 3H intracellular distribution and release were studied in vitro in the rat hypothalamus. 2. A III (1 microM) decreased total, neuronal and non-neuronal 3H-NA uptake
K S Lin et al.
The American journal of physiology, 272(5 Pt 2), H2204-H2210 (1997-05-01)
We evaluated the role of endogenous angiotensin II and III (ANG II and ANG III) at the rostral nucleus reticularis ventrolateralis (NRVL) in the modulation of baroreceptor reflex (BRR) response and the subtype of angiotensin receptors involved in this process.
K L Clark et al.
Journal of cardiovascular pharmacology, 19(4), 515-524 (1992-04-01)
Experiments were performed in anesthetized cats to compare the characteristics of angiotensin receptors in the renal and mesenteric vascular beds. Injection of either angiotensin II (Ang II; 0.3-30 ng) or angiotensin III (Ang III; 0.3-30 ng) directly into the superior
E L Bravo et al.
The Journal of clinical endocrinology and metabolism, 40(3), 530-533 (1975-03-01)
The vascular and steroidogenic responses to (1-Sarcosine, 8-Isoleucine)-, and to (1-Des (Aspartic acid), 8-Isoleucine) angiotensin II were compared in bilaterally nephrectomized, ACTH-suppressed dogs receiving constant infusions of angiotensin II. Aldosterone secretion rate was significantly inhibited by pretreatment with 200 ng/Kg/min
M J Robertson et al.
British journal of pharmacology, 106(1), 166-172 (1992-05-01)
1. Experiments were performed with peptidase inhibitors on rabbit aortic strip preparations, to determine whether endogenous peptidase activity can influence the potency estimates for angiotensin receptor agonists and antagonists in this tissue. 2. Angiotensin II (A II) and angiotensin III

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service