Skip to Content
Merck
All Photos(1)

Key Documents

UC454

Sigma-Aldrich

3-Cyano-7-methoxycoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7NO3
CAS Number:
Molecular Weight:
201.18
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

pale yellow

mp

210-212 °C

storage temp.

2-8°C

SMILES string

COc1ccc2C=C(C#N)C(=O)Oc2c1

InChI

1S/C11H7NO3/c1-14-9-3-2-7-4-8(6-12)11(13)15-10(7)5-9/h2-5H,1H3

InChI key

GGAFPIBJPYCFGR-UHFFFAOYSA-N

Application

3-Cyano-7-methoxycoumarin can be used as a fluorophore in cytochrome P450 (CYP)-based fluorescent assays.

Biochem/physiol Actions

Fluorescent CYP1A1 &1A2 substrate

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service