Skip to Content
Merck
All Photos(2)

Key Documents

SCC603

Sigma-Aldrich

3dGRO® Human Lung Organoids (XDO.344)

Human

Synonym(s):

Lung PDO;Lung Tumor Organoid;Airway Organoid;Pulmonary Organoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
NACRES:
NA.28
Pricing and availability is not currently available.

Product Name

3dGRO® Human Lung Organoids (XDO.344),

biological source

human

Quality Level

packaging

vial of ≥ 1500 organoids/vial mg

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | stem cell: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

  • Each vial contains >1500 organoids.
  • Mycoplasma: Negative
  • Genotyped by STR analysis: Pass
  • Human species verified
  • Organoids are negative for HPV-16, HPV-18, Hepatitis A, B, C, and HIV-1 & 2 viruses by PCR.
  • Cell viability test: Pass

Patient-derived lung organoids (lung PDOs) are adult tissue derived in vitro cultured “mini-lungs” that accurately replicate the histological and functional aspects of in vivo lung tissue. Lung organoids have been shown to contain all 6 major lung epithelial cell types including AEC2s, AEC1s, basal cells, club cells, goblet cells and ciliated cells. These 3D organoid models have become attractive alternatives to traditional PDX and animal lung models to study pulmonary diseases including COPD, cystic fibrosis, SARS-CoV-2, viral infections, asthma, and lung cancer. The 3dGRO® Human Lung Organoids are high quality cryopreserved cell lines derived from patient-derived xenografts, primary tumors or metastatic patient tissues from various regions of the human lung (acinar, papillary, solid, squamous, mucinous). These lung organoids are highly characterized and express lung airway markers including EPCAM, cytokeratin (CK5, CK7), acetyl-α-tubulin, surfactant proteins (SFTPB/SFTPC), TTF1, P63 and Muc5AC.

Cell Line Characteristics

Sex: Male
Age: 46
Organ: Lung (PDX- Acinar/Solid)
Disease: Adenocarcinoma (Driver Mutations: KRAS G12C)

Features and Benefits

Cryopreserved Male Adenocarcinoma Lung Organoids (Acinar/Solid)

Storage and Stability

3dGRO® human lung organoids should be stored in liquid nitrogen.

Other Notes

3dGRO® is a trademark of KGaA, Darmstadt, Germany.3dGRO® organoids were derived utilizing HUB Organoid Technology.The purchaser of this product shall agree to HUB′s terms of use, which shall be separately acknowledged and accepted by such purchaser, prior to transfer of this product to purchaser.

Legal Information

3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore our cryopreserved human lung organoid biobank from NSCLC tumor biopsies for lung cancer, SARS-CoV2, COPD and pulmonary disease research. Lung organoid 3D cell culture protocol available.

Related Content

Explore our portfolio of 3D cell culture tools and technologies for any 3D cell line or application and discover organoids, hydrogels, ULA, microwell, or hydrogel plates, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service