Skip to Content
Merck
All Photos(1)

Key Documents

TMO569-0020

Nalgene® vacuum filtration system

filter capacity 500 mL, polyethersulfone, pore size 0.2 μm, case of 12 ea

Synonym(s):

moisture extraction device, sample protection unit

Sign Into View Organizational & Contract Pricing

Select a Size

12 EACH
$328.00

$328.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
12 EACH
$328.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

$328.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polyethersulfone
polyethersulfone membrane

packaging

case of 12 ea

manufacturer/tradename

Nalgene 569-0020

filter capacity

500 mL

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Increase filtration efficiency by using the Nalgene® Rapid-Flow disposable filter units featuring the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination when filtering cell culture media, serum, additives and buffers. With a graduated upper chamber and Polyethersulfone (PES) membrane this filter unit offer higher aqueous flow rates; lower extractables (no wetting agent); lower protein binding; and greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service