Key Documents
TMO382019-2000
Nalgene® Certified Clean bottles
clear PETG, volume 2000 mL, case of 12 ea
Synonym(s):
PETG bottle, certified nalgene bottle
Select a Size
12 EACH
$610.00
$610.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
12 EACH
$610.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$610.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
clear PETG
packaging
case of 12 ea
manufacturer/tradename
Nalgene 382019-2000
capacity
2000 mL
volume
2000 mL
General description
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Features and Benefits
  • Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)