Key Documents
TMO362004-9050
Nalgene® diagnostic bottle without cap
amber high-density polyethylene, volume 15 mL, case of 2000 ea
Synonym(s):
PPCO bottles, small volume bottle
Select a Size
2000 EACH
$1,360.00
$1,360.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
2000 EACH
$1,360.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$1,360.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
amber high-density polyethylene
packaging
case of 2000 ea
manufacturer/tradename
Nalgene 362004-9050
capacity
15 mL
volume
15 mL
General description
Safely store and ship light-sensitive diagnostic samples and valuable reagents in small volumes with Thermo Scientific Nalgene Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque amber high-density polyethylene bottles are for use with light sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)