Key Documents
TMO278616
Nunc® 1mL 96 DeepWell(TM) plate Well Cap Mat
chemically resistant, sterile, case of 25 ea
Synonym(s):
plate cover, plate cover mat
Select a Size
25 EACH
$564.00
$564.00
Estimated to ship onMarch 28, 2025
Select a Size
Change View
25 EACH
$564.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
$564.00
Estimated to ship onMarch 28, 2025
sterility
sterile
packaging
case of 25 ea
manufacturer/tradename
Nalge-Nunc 278616
General description
Protect plate contents during short- and long-term storage of chemicals or compounds by using the flexible Thermo Scientific Nunc 96 Well Caps with 96 indentations specifically desgined for the 96 wells in Thermo Scientific Nunc 1.0mL Polystyrene DeepWell Plates (278605 and 278606).
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)