Skip to Content
Merck
All Photos(1)

Key Documents

TMO157150

Nunclon® cell culture dishes

diam. 150 mm, with lid, Nunclon® Delta treated, case of 120 ea

Synonym(s):

cell culture dish, cell culture treateddish

Sign Into View Organizational & Contract Pricing

Select a Size

120 EACH
$726.00

$726.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
120 EACH
$726.00

About This Item

UNSPSC Code:
41121812
NACRES:
NB.15

$726.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polystyrene dishes with lids

sterility

sterile; γ-irradiated

packaging

case of 120 ea

manufacturer/tradename

Nunc 157150

diam.

150 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Large range available, treated for cell culture but with many other uses
  • Optically clear and very flat for use in microscope
  • Non-toxic
  • Certified surface treatment for optimal cell attachment and growth

Application

Nunclon® cell culture dishes are suitable for a wide range of cellculture routines.

Features and Benefits

  • Nunclon® Delta surface treated for maximum adhesion of a wide variety of cells
  • Sterile and non-pyrogenic
  • Stable stacking and easier handling due to stacking rings at the top and bottom
  • For gas exchange, the plates are available with or without vents
  • Grid for cloning experiments and determination of plating efficiency

Legal Information

Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service