Key Documents
T3043
Nalgene® centrifuge tubes, Oak Ridge Style 3138
nominal capacity 10 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes, Nalgene tubes
Select a Size
10 EA
$242.00
$242.00
Estimated to ship onMay 29, 2025
Select a Size
Change View
10 EA
$242.00
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
$242.00
Estimated to ship onMay 29, 2025
material
cap (Sealing cap)
round bottom tube
transparent polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3138-0010
nominal capacity
10 mL
General description
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.
Packaging
With sealing cap
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)