Skip to Content
Merck
All Photos(1)

Documents

T1818

Sigma-Aldrich

Greiner centrifuge tubes

centrifuge tube, 15 mL, 17x120 mm, conical (V) bottom, w/ graduation, I.D. field, w/ rack

Synonym(s):

centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, Greiner centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.11

material

cap
conical bottom tube
polypropylene tube

sterility

sterile

feature

RCF 3,500 × g
graduations, ID field, w/ rack

packaging

pack of 50
case of 500

manufacturer/tradename

Greiner 188261-N

capacity

15 mL

diam. × L

17 mm × 120 mm

color

blue cap
colorless tube

Looking for similar products? Visit Product Comparison Guide

General description

Greiner centrifuge tubes
High quality, crystal-clear polypropylene centrifuge tubes. Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Complete workflow for cannabis flower pesticide testing using LC-MS/MS and GC-MS/MS ensures product safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service