SYNP191001
Synthware™ class A single volume pipet
bottle capacity 1 mL, borosilicate glass 3.3
Synonym(s):
Synthware P191001, Pasteur pipette
About This Item
Recommended Products
material
borosilicate glass 3.3
packaging
pack of 1 ea
manufacturer/tradename
Kemtech P191001
bottle capacity
1 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service