SYNF710010YH
Synthware™ glass graduated cylinder
borosilicate glass 3.3, round base, capacity 10 mL
Synonym(s):
Synthware F710010YH
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3
round base
feature
class A
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F710010YH
capacity
10 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Graduated Cylinder, Round Base (Class A) 10mL, Boro 3.3
Application
kemetch; synthware; GRADUATED CYLINGER; class a
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service