SRP5335
PP2C-α, active, GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
FLJ42306, MGC9201, PP2C-ALPHA, PP2CA, PPM1A
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
960-1440 nmol/min·mg
mol wt
~105 kDa
NCBI accession no.
application(s)
cell analysis
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PPM1A(5494)
General description
Protein phosphatase 2C-α (PP2C-α), also called protein phosphatase, Mg2+/Mn2+ dependent 1A (PPM1A), is a member of the family of Ser/Thr protein phosphatases and is composed of a catalytic subunit and a constant regulatory subunit. PP2C-α is highly expressed in heart and skeletal muscle. The gene encoding this protein is localized on human chromosome 4q12.
Biochem/physiol Actions
Protein phosphatase 2C-α (PP2C-α) or PPM1A is known to be negative regulators of cell stress response pathways. It has been reported to negatively regulate TGF-β signaling pathway and cell cycle. PP2C-α has been shown to dephosphorylate cyclin-dependent kinases (CDKs) and it may be involved in cell cycle control. It modulates cell migration and invasion. Overexpression of PP2C-α is reported to cause cell-growth arrest or cell death.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Preparation Note
After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Integrated analysis of expression and genome alteration reveals putative amplified target genes in esophageal cancer.
Oncology Reports (2007)
The cloning expression and tissue distribution of human PP2C?1
Febs Letters (1998)
PPM1A Methylation Is Associated With Vascular Recurrence in Aspirin-Treated Patients.
Stroke (2016)
Loss of PPM1A expression enhances invasion and the epithelial-to-mesenchymal transition in bladder cancer by activating the TGF-?/Smad signaling pathway.
Oncotarget (2014)
Dephosphorylation of Human Cyclin-dependent Kinases by Protein Phosphatase Type 2Ca and ?2 Isoforms*
The Journal of Biological Chemistry (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service