Skip to Content
Merck
All Photos(1)

Key Documents

14-595-M

Sigma-Aldrich

PP1α Protein, 10 µg

Recombinant, full length, human PP1α, for use in Phosphatase Assays.

Synonym(s):

PPP1C, Phosphoprotein phosphatase 1, Phosphorylase phosphatase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

form

solution

shelf life

6 mo.

mol wt

Mw 37.6 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

solubility

water: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PPP1CA(5499)

General description

Research area: Neuroscience
Protein Phosphatase 1 (PP1) is one of the key serine/threonine-specific protein phosphatases.

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: PP1α Target Sub-Family: Ser/Thr phosphatase

Protein Phosphatase 1 (PP1) has been associated with a wide range of biological processes, including synaptic plasticity, cell cycle progression, dopaminergic neurotransmission, contraction, and glycogen metabolism, maintaining microtubule stability and the dephosphorylation of tau. Furthermore, PP-1 plays a vital role in synaptic development, plasticity, signaling, neurite outgrowth, synapse formation, ion channel function, and neurotransmission. Changes in PP-1 signaling within dendritic spines have been linked to various neurological and psychiatric disorders.

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-595M when ordering the 250 µg size.

Quality

routinely evaluated by dephosphorylation of 100μM DiFMUP

Physical form

Heparin agarose followed by Q agarose

Storage and Stability

6 months at -20°C from date of shipment

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karl Foley et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(14), 3040-3050 (2021-04-09)
Protein phosphatases, by counteracting protein kinases, regulate the reversible phosphorylation of many substrates involved in synaptic plasticity, a cellular model for learning and memory. A prominent phosphatase regulating synaptic plasticity and neurologic disorders is the serine/threonine protein phosphatase 1 (PP1).
The role of protein phosphatase-1 in the modulation of synaptic and structural plasticity.
Munton, Richard P, et al.
Febs Letters, 567, 121-128 (2004)
Targeting phosphatases as the next generation of disease modifying therapeutics for Parkinson?s disease
Braithwaite SP, et al.
Neurochemistry International, 61, 899-906 (2012)
Modulation of dendritic spines by protein phosphatase-1
Advances in Pharmacology, 90.0, 117-144 (2021)
Alphonse Garcia et al.
Biochimie, 85(8), 721-726 (2003-10-31)
The reversible phosphorylation of proteins controlled by protein kinases and protein phosphatases is a major mechanism that regulates a wide variety of cellular processes. In contrast to C. elegans, recent studies in mammalian cells have highlighted a major role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service