Skip to Content
Merck
All Photos(2)

Documents

SRP5249

Sigma-Aldrich

PDHK3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PDK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

10.2-13.8 nmol/min·mg

mol wt

~69 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDK3(5165)

Biochem/physiol Actions

PDHK3 or Pyruvate Dehydrogenase Kinase 3 is a member of the PDHK family that phosphorylate and inactivate the Pyruvate Dehydrogenase (PDH). PDH is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. HIF-1 induces PDHK3 expression leading to inhibition of mitochondrial respiration. Forced expression of PDHK3 in cancer cells results in increased lactic acid accumulation and drugs resistance, whereas knocking down PDHK3 inhibits hypoxia-induced cytoplasmic glycolysis and cell survival.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Wun Lu et al.
The Journal of biological chemistry, 283(42), 28106-28114 (2008-08-23)
The switch of cellular metabolism from mitochondrial respiration to glycolysis is the hallmark of cancer cells and associated with tumor malignancy. However, the mechanism of this metabolic switch remains largely unknown. Herein, we reported that hypoxia-inducible factor-1 (HIF-1) induced pyruvate
R Gudi et al.
The Journal of biological chemistry, 270(48), 28989-28994 (1995-12-01)
Recent evidence from this laboratory indicates that at least two isoenzymic forms of pyruvate dehydrogenase kinase (PDK1 and PDK2) may be involved in the regulation of enzymatic activity of mammalian pyruvate dehydrogenase complex by phosphorylation (Popov, K.M., Kedishvili, N.Y., Zhao

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service