Skip to Content
Merck
All Photos(1)

Documents

14-452-M

Sigma-Aldrich

PDK1 Protein, active, 10 µg

Active, N-terminal, His6-tagged, recombinant human PDK1, amino acids 52-end. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.26

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 59 kDa

purified by

(Ni2+/NTA-agarose)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

accession no.

 (FUNCTION: SwissProt: O15530 # Phosphorylates and activates not only PKB/AKT, but also PKA, PKC-zeta, RPS6KA1 and RPS6KB1. May play a general role in signaling processes and in development (By similarity). Isoform 3 is catalytically inactive. )
NM_002613.3

NCBI accession no.

UniProt accession no.

Gene Information

human ... PDPK1(5170)

General description

N-terminal, His6-tagged, recombinant human PDK1, amino acids 52-end.

Biochem/physiol Actions

Protein Target: PDK1
Target Sub-Family: AGC

Packaging

Also available in 10 μg size and in bulk
Also available in 250 µg size --call for pricing and availability and reference catalog number 14-452M when ordering the 250 µg size.

Quality

Routinely evaluated by phosphorylation of PDKtide

Target description

PDK1

Physical form

50mM Tris-HCl, pH 7.5, 0.15M NaCl, 0.1mM EGTA, 0.03% Brij-35, 0.1% 2-mercaptoethanol, 1mM benzamidine, 200μM PMSF and 5% glycerol
Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service