Skip to Content
Merck
All Photos(1)

Key Documents

SRP3291

Sigma-Aldrich

NESFATIN-1 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

none

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$437.00

$437.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
100 μG
$437.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

$437.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

9.7 kDa

packaging

pkg of 100 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NUCB2(4925)

General description

Originally identified as a hypothalamic neuropeptide, Nesfatin-1 is also expressed in other areas of the brain, and in pancreatic islets β-cells, gastric endocrine cells and adipocytes.  Nesfatin-1 suppresses food intake and can regulate energy metabolism in a Leptin independent manner.

Biochem/physiol Actions

Nesfatin-1 is a metabolic polypeptide encoded in the N-terminal region of the precursor protein, Nucleobindin2 (NUCB2). Recombinant human Nesfatin-1 is a 9.7 kDa protein containing 82 amino acid residues.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 6.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Stengel et al.
Regulatory peptides, 163(1-3), 18-23 (2010-06-29)
Nesfatin-1 is an 82 amino acid peptide recently discovered in the brain which is derived from nucleobindin2 (NUCB2), a protein that is highly conserved across mammalian species. Nesfatin-1 has received much attention over the past two years due to its
Yusaku Mori et al.
Endocrine connections, 8(5), 536-546 (2019-04-03)
Nesfatin-1 is a novel anorexic peptide hormone that also exerts cardiovascular protective effects in rodent models. However, nesfatin-1 treatment at high doses also exerts vasopressor effects, which potentially limits its therapeutic application. Here, we evaluated the vasoprotective and vasopressor effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service