Skip to Content
Merck

Skip To

39294

Fluo 3-AM solution

1 mM in DMSO, ≥99.0% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mL
Check Cart for Availability
$639.00

About This Item

Empirical Formula (Hill Notation):
C51H50Cl2N2O23
CAS Number:
Molecular Weight:
1129.85
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.32

$639.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (TLC)

form

liquid

concentration

1 mM in DMSO

fluorescence

λex 506 nm; λem 525.0 nm±5.0 nm in 0.1 M Tris pH 8.0, esterase

storage temp.

−20°C

SMILES string

Clc1[c](cc2[o]c3c(c(c2c1)c4cc(c(cc4)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOc5c(ccc(c5)C)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)cc(c(c3)OCOC(=O)C)Cl)=O

InChI

1S/C51H50Cl2N2O23/c1-28-7-9-39(54(19-47(62)74-24-69-30(3)57)20-48(63)75-25-70-31(4)58)45(13-28)66-11-12-67-46-14-34(8-10-40(46)55(21-49(64)76-26-71-32(5)59)22-50(65)77-27-72-33(6)60)51-35-15-37(52)41(61)17-42(35)78-43-18-44(38(53)16-36(43)51)73-23-68-29(2

InChI key

PTPUOMXKXCCSEN-UHFFFAOYSA-N

General description

Cell permeable fluorescent indicator of intracellular Ca2+; non-fluorescent until it is hydrolyzed intracellularly and/or in the presence of Ca2+; lower binding affinity allows measurement of higher peaks of Ca2+ transients than with Fura 2.

Application

suitable as fluorescent Ca sensitive probe (after cleavage of the AM ester)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
343242-M34490647989
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

solid

form

liquid

form

solid

assay

≥99.0% (TLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95.0% (HPLC)

concentration

1 mM in DMSO

concentration

-

concentration

-

concentration

-

fluorescence

λex 506 nm; λem 525.0 nm±5.0 nm in 0.1 M Tris pH 8.0, esterase

fluorescence

-

fluorescence

λex 335 nm (high), λex 362 nm (low), λem 505 nm (high), λem 512 nm (low)

fluorescence

λex 355 nm; λem 495 nm in 0.1 M Tris pH 8.0, esterase; 10 mM Ca2+

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F

flash_point_c

87 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Shiwen Han et al.
Scientific reports, 7(1), 7222-7222 (2017-08-05)
Glycyrrhizic acid (GA), the major bioactive triterpene glycoside of glycyrrhiza, has been shown to possess a wide range of pharmacological properties, including anti-inflammatory and anti-viral properties. However, few studies have examined the anti-allergic activity and exact mechanism of action of
Suting Li et al.
Cell death & disease, 11(4), 279-279 (2020-04-26)
Multiple vaginal delivery (MVD) is an important factor for pelvic floor muscle (PFM) function decline and pelvic floor dysfunction (PFD). PFD is common in middle-aged and elderly women, but its pathogenesis is not clear. In this study, we found that
Shrawan Kumar Mishra et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1864(2), 158-167 (2018-11-26)
Human GLTP on chromosome 12 (locus 12q24.11) encodes a 24 kD amphitropic lipid transfer protein (GLTP) that mediates glycosphingolipid (GSL) intermembrane trafficking and regulates GSL homeostatic levels within cells. Herein, we provide evidence that GLTP overexpression inhibits the growth of



Global Trade Item Number

SKUGTIN
39294-1ML04061831981766

Questions

Reviews

No rating value

Active Filters