Skip to Content
Merck
All Photos(1)

Key Documents

SRP3154

Sigma-Aldrich

sFRP-1 human

recombinant, expressed in HeLa cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture

Synonym(s):

SARP-2, Secreted Apoptosis Related Protein-2, Soluble Frizzled Related Protein-1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$522.00

$522.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
$522.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

$522.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in HeLa cells

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

potency

0.3-0.5 μg/mL ED50

mol wt

31.0-36.0 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SFRP1(6422)

General description

The gene SFRP1 (secreted frizzled related protein 1) is mapped to human chromosome 8p22–p12, a region frequently deleted in breast cancer. It is highly expressed in normal breast tissue.[1][2]

Biochem/physiol Actions

Secreted Frizzled-related protein 1 (SFRP1) is a secreted inhibitor of Wnt signaling pathway. DNA damage or oxidative stress causes cellular senescence, which leads to overexpression of sfrp1. This protein induces senescence phenotypes via the inactivation of Wnt signaling and activation of the retinoblastoma (Rb) pathway.[3] Mutations or aberrant methylation in this gene impair the induction of senescence in several cancers, including breast cancer, bladder cancer, and colorectal tumorigenesis.[3][4][5][6]

Sequence

SEYDYVSFQS DIGPYQSGRF YTKPPQCVDI PADLRLCHNV GYKKMVLPNL LEHETMAEVK QQASSWVPLL NKNCHAGTQV FLCSLFAPVC LDRPIYPCRW LCEAVRDSCE PVMQFFGFYW PEMLKCDKFP EGDVCIAMTP PNATEASKPQ GTTVCPPCDN ELKSEAIIEH LCASEFALRM KIKEVKKENG DKKIVPKKKK PLKLGPIKKK DLKKLVLYLK NGADCPCHQL DNLSHHFLIM GRKVKSQYLL TAIHKWDKKN KEFKNFMKKM KNHECPTFQS VFK

Physical form

Lyophilized from 10 mM Sodium Phosphate pH 7.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

WNT pathway and mammary carcinogenesis: loss of expression of candidate tumor suppressor gene SFRP1 in most invasive carcinomas except of the medullary type.
Ugolini F
Oncogene, 20, 5810-5817 (2001)
A M Goodwin et al.
Angiogenesis, 5(1-2), 1-9 (2003-01-29)
The Wnt signaling pathway regulates normal development as well as a variety of pathologies. Studies of the Wnt pathway have focused largely on very early development and on tumorigenesis. Recent observations point to a role for Wnt signaling in vessel
Methylation-associated silencing of SFRP1 with an 8p11-12 amplification inhibits canonical and non-canonical WNT pathways in breast cancers.
Yang ZQ
International Journal of Cancer. Journal International Du Cancer, 125, 1613-1621 (2009)
J Veeck et al.
Oncogene, 25(24), 3479-3488 (2006-02-02)
The canonical Wnt signalling pathway plays a key role during embryogenesis and defects in this pathway have been implicated in the pathogenesis of various types of tumours, including breast cancer. The gene for secreted frizzled-related protein 1 (SFRP1) encodes a
David J Elzi et al.
Molecular and cellular biology, 32(21), 4388-4399 (2012-08-29)
Cellular senescence has emerged as a critical tumor suppressive mechanism in recent years, but relatively little is known about how senescence occurs. Here, we report that secreted Frizzled-related protein 1 (SFRP1), a secreted antagonist of Wnt signaling, is oversecreted upon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service