Skip to Content
Merck
All Photos(1)

Key Documents

SRP3107

Sigma-Aldrich

Lin28-TAT human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC), suitable for cell culture

Synonym(s):

CSDD1, LIN28A, ZCCHC1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$479.00

$479.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
25 μG
$479.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$479.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (HPLC)
≥90% (SDS-PAGE)

form

lyophilized

mol wt

24.4 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LIN28A(79727)

General description

Lin28 is a RNA-binding protein that belongs to a diverse family of structurally-related transcription factors. Lin28 is found abundantly in embryonic stem cells (ESCs), and to a lesser extent in placenta and testis. Lin28 has been shown to block let-7 microRNA processing and maturation, a necessary step in the differentiation of stem cells and certain cancer cell lines. Together with Sox2, Oct4, and Nanog, Lin28 can induce the reprogramming of primary human fibroblasts to a pluripotent state. Lin28 and other regulatory proteins can be introduced into cells by DNA transfection, viral infection, or microinjection. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing proteins into primary as well as transformed cells. Recombinant human Lin28-TAT is a 24.4 kDa protein containing 222 amino acid residues, including 13- residue C-terminal TAT peptide.

Biochem/physiol Actions

Lin28 is a RNA-binding protein that belongs to a diverse family of structurally-related transcription factors. Recombinant human Lin28-TAT is a 24.4 kDa protein containing 222 amino acid residues, including 13- residue C-terminal TAT peptide.

Sequence

GPSVSNQQFA GGCAKAAEEA PEEAPEDAAR AADEPQLLHG AGICKWFNVR MGFGFLSMTA RAGVALDPPV DVFVHQSKLH MEGFRSLKEG EAVEFTFKKS AKGLESIRVT GPGGVFCIGS ERRPKGKSMQ KRRSKGDRCY NCGGLDHHAK ECKLPPQPKK CHFCQSISHM VASCPLKAQQ GPSAQGKPTY FREEEEEIHS PTLLPEAQNG GYGRKKRRQR RR

Physical form

Lyophilized from 10 mM Sodium Citrate, pH 3.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingsen Xu et al.
RNA (New York, N.Y.), 15(3), 357-361 (2009-01-17)
Lin28 is highly expressed in human and mouse embryonic stem (ES) cells. Here, we show that in mouse ES cells, specific repression of Lin28 results in decreased cell proliferation, while overexpression of Lin28 accelerates cell proliferation. Further, Lin28 associates specifically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service